Search
Search
| Gene Symbol: BRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605512.3 | XP_005605569.2 | 603 | 75 | ||
| XM_005605513.3 | XP_005605570.2 | 601 | 75 | |||
| XM_023628561.1 | XP_023484329.1 | 917 | 75 | |||
| XM_023628562.1 | XP_023484330.1 | 918 | 75 | |||
| XM_023628563.1 | XP_023484331.1 | 795 | 75 | |||
| XM_023628564.1 | XP_023484332.1 | 918 | 75 | |||
| XM_023628565.1 | XP_023484333.1 | 730 | 75 | |||
| XM_023628566.1 | XP_023484334.1 | 524 | 75 | |||
Set Membership: |
|