Search
Search

PHF24| Gene Symbol: PHF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014735873.2 | XP_014591359.1 | 2160 | 91 | ||
| XM_023629587.1 | XP_023485355.1 | 798 | 91 | |||
| XM_023629588.1 | XP_023485356.1 | 2335 | 91 | |||
| XM_023629589.1 | XP_023485357.1 | 1034 | 91 | |||