Search
Search
| Gene Symbol: TLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605599.3 | XP_005605656.2 | 3481 | 59 | ||
| XM_014735893.2 | XP_014591379.2 | 3532 | 59 | |||
| XM_014735894.2 | XP_014591380.2 | 3531 | 59 | |||
| XM_023629651.1 | XP_023485419.1 | 3119 | 59 | |||
Set Membership: |
|