Search
Search

RNF38| Gene Symbol: RNF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605623.3 | XP_005605680.1 | 569 | 84 | ||
| XM_005605625.3 | XP_005605682.1 | 1020 | 84 | |||
| XM_023629669.1 | XP_023485437.1 | 617 | 84 | |||
| XM_023629670.1 | XP_023485438.1 | 631 | 84 | |||
| XM_023629671.1 | XP_023485439.1 | 1171 | 84 | |||
| XM_023629672.1 | XP_023485440.1 | 1069 | 84 | |||
| XM_023629673.1 | XP_023485441.1 | 257 | 84 | |||