Search
Search

RNF38| Gene Symbol: RNF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605623.3 | XP_005605680.1 | 738 | 67 | ||
| XM_005605625.3 | XP_005605682.1 | 1189 | 67 | |||
| XM_023629669.1 | XP_023485437.1 | 786 | 67 | |||
| XM_023629670.1 | XP_023485438.1 | 800 | 67 | |||
| XM_023629671.1 | XP_023485439.1 | 1340 | 67 | |||
| XM_023629672.1 | XP_023485440.1 | 1238 | 67 | |||
| XM_023629673.1 | XP_023485441.1 | 426 | 67 | |||
Set Membership: |
|