Search
Search

TDRD7| Gene Symbol: TDRD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629700.1 | XP_023485468.1 | 2612 | 70 | ||
| XM_023629701.1 | XP_023485469.1 | 2621 | 70 | |||
| XM_023629702.1 | XP_023485470.1 | 2555 | 70 | |||
Set Membership: |
|