Search
Search
| Gene Symbol: INVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605658.3 | XP_005605715.2 | 2982 | 63 | ||
| XM_023629745.1 | XP_023485513.1 | 2834 | 63 | |||
| XM_023629746.1 | XP_023485514.1 | 2641 | 63 | |||
Set Membership: |
|