Search
Search
| Gene Symbol: ELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014735945.2 | XP_014591431.2 | 3659 | 101 | ||
| XM_023629846.1 | XP_023485614.1 | 3844 | 101 | |||
| XM_023629847.1 | XP_023485615.1 | 3844 | 101 | |||
| XM_023629848.1 | XP_023485616.1 | 3990 | 101 | |||
| GenBank mRNA | JL639471.1 | - | 5 - 6 | 440 | 101 | |
Set Membership: |
|