Search
Search

TMEM245| Gene Symbol: TMEM245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629850.1 | XP_023485618.1 | 638 | 58 | ||
| XM_023629851.1 | XP_023485619.1 | 638 | 58 | |||
| XM_023629852.1 | XP_023485620.1 | 640 | 58 | |||
| XM_023629853.1 | XP_023485621.1 | 639 | 58 | |||
Set Membership: |
|