Search
Search

COL27A1| Gene Symbol: COL27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628721.1 | XP_023484489.1 | 2033 | 91 | ||
| XM_023628722.1 | XP_023484490.1 | 2251 | 91 | |||
| XM_023628725.1 | XP_023484493.1 | 2033 | 91 | |||
| XM_023628726.1 | XP_023484494.1 | 2033 | 91 | |||
Set Membership: |
|