Search
Search

ESPL1| Gene Symbol: ESPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611226.2 | XP_005611283.2 | 3063 | 72 | ||
| XM_005611227.2 | XP_005611284.2 | 2982 | 72 | |||
| XM_023643649.1 | XP_023499417.1 | 1278 | 72 | |||
Set Membership: |
|