Search
Search

RAPGEF1| Gene Symbol: RAPGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499272.6 | XP_001499322.2 | 1194 | 74 | ||
| XM_005605889.3 | XP_005605946.1 | 2004 | 74 | |||
| XM_023629173.1 | XP_023484941.1 | 1287 | 74 | |||
| XM_023629174.1 | XP_023484942.1 | 1304 | 74 | |||
| XM_023629176.1 | XP_023484944.1 | 1173 | 74 | |||
| XM_023629177.1 | XP_023484945.1 | 1900 | 74 | |||
| XM_023629178.1 | XP_023484946.1 | 1191 | 74 | |||
| XM_023629179.1 | XP_023484947.1 | 1287 | 74 | |||
| XM_023629180.1 | XP_023484948.1 | 1294 | 74 | |||
Set Membership: |
|