Search
Search

DYRK1A| Gene Symbol: DYRK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736373.2 | XP_014591859.2 | 1850 | 103 | ||
| XM_023630144.1 | XP_023485912.1 | 1877 | 103 | |||
| XM_023630145.1 | XP_023485913.1 | 1766 | 103 | |||
| XM_023630146.1 | XP_023485914.1 | 1970 | 103 | |||
| XM_023630147.1 | XP_023485915.1 | 1769 | 103 | |||
| XM_023630148.1 | XP_023485916.1 | 1943 | 103 | |||
Set Membership: |
|