Search
Search

ADARB1| Gene Symbol: ADARB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489891.5 | XP_001489941.4 | 2013 | 81 | ||
| XM_005606186.3 | XP_005606243.2 | 2019 | 81 | |||
| XM_014736437.2 | XP_014591923.1 | 2892 | 81 | |||
| XM_023630298.1 | XP_023486066.1 | 1270 | 81 | |||
| XM_023630299.1 | XP_023486067.1 | 1217 | 81 | |||
Set Membership: |
|