Search
Search

COL6A2| Gene Symbol: COL6A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630335.1 | XP_023486103.1 | 1838 | 60 | ||
| XM_023630336.1 | XP_023486104.1 | 1827 | 60 | |||
| XM_023630337.1 | XP_023486105.1 | 1838 | 60 | |||
Set Membership: |
|