Search
Search

PRMT2| Gene Symbol: PRMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630373.1 | XP_023486141.1 | 1495 | 91 | ||
| XM_023630374.1 | XP_023486142.1 | 1788 | 91 | |||
| XM_023630375.1 | XP_023486143.1 | 1353 | 91 | |||
| XM_023630376.1 | XP_023486144.1 | 1499 | 91 | |||
| XM_023630377.1 | XP_023486145.1 | 1492 | 91 | |||
| XM_023630378.1 | XP_023486146.1 | 1784 | 91 | |||
| XM_023630380.1 | XP_023486148.1 | 1495 | 91 | |||
| GenBank mRNA | JL634590.1 | - | 1 - 2 | 434 | 91 | |
Set Membership: |
|