Search
Search

PLPP5| Gene Symbol: PLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606319.3 | XP_005606376.2 | 828 | 78 | ||
| XM_023630619.1 | XP_023486387.1 | 420 | 78 | |||
| XM_023630620.1 | XP_023486388.1 | 705 | 78 | |||
Set Membership: |
|