Search
Search

C27H4orf47| Gene Symbol: C27H4orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916303.4 | XP_001916338.1 | 455 | 99 | ||
| XM_005606379.3 | XP_005606436.1 | 483 | 99 | |||
| XM_023630945.1 | XP_023486713.1 | 393 | 99 | |||