Search
Search

KCNC2| Gene Symbol: KCNC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023631660.1 | XP_023487428.1 | 1313 | 89 | ||
| XM_023631661.1 | XP_023487429.1 | 841 | 89 | |||
| XM_023631662.1 | XP_023487430.1 | 1313 | 89 | |||
Set Membership: |
|