Search
Search

SREBF2| Gene Symbol: SREBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606691.3 | XP_005606748.1 | 2660 | 87 | ||
| XM_023631221.1 | XP_023486989.1 | 2886 | 87 | |||
| XM_023631222.1 | XP_023486990.1 | 2766 | 87 | |||