Search
Search
| Gene Symbol: EPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606850.3 | XP_005606907.1 | 589 | 75 | ||
| XM_023632004.1 | XP_023487772.1 | 761 | 75 | |||
| XM_023632005.1 | XP_023487773.1 | 760 | 75 | |||
| XM_023632006.1 | XP_023487774.1 | 6191 | 75 | |||
Set Membership: |
|