Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ec06988412_m1
See other KIAA1217 GE Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
-
Chromosome Location:
Chr.29: 13104689 - 13398565 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID
Ec06988412_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_001495617.5
XP_001495667.1
2228
81
XM_003364351.4
XP_003364399.1
2230
81
XM_014737039.2
XP_014592525.1
2311
81
XM_014737040.2
XP_014592526.1
2314
81
XM_014737041.2
XP_014592527.1
2310
81
XM_014737042.2
XP_014592528.1
2229
81
XM_014737043.2
XP_014592529.1
2206
81
XM_014737044.2
XP_014592530.1
2311
81
XM_014737045.2
XP_014592531.1
2205
81
XM_014737046.2
XP_014592532.1
2310
81
XM_014737048.2
XP_014592534.1
2124
81
XM_014737049.2
XP_014592535.1
6037
81
XM_014737050.2
XP_014592536.1
5066
81
XM_014737051.2
XP_014592537.1
3371
81
XM_014737052.2
XP_014592538.1
1604
81
XM_014737053.2
XP_014592539.1
3821
81
XM_014737054.2
XP_014592540.1
2314
81
XM_014737055.2
XP_014592541.1
2314
81
XM_014737056.2
XP_014592542.1
2125
81
XM_014737057.2
XP_014592543.1
2205
81
XM_023632151.1
XP_023487919.1
4286
81
XM_023632152.1
XP_023487920.1
4538
81
XM_023632154.1
XP_023487922.1
2317
81
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Equus caballus
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline