Search
Search

TRDMT1| Gene Symbol: TRDMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632064.1 | XP_023487832.1 | 828 | 85 | ||
| XM_023632066.1 | XP_023487834.1 | 827 | 85 | |||
| XM_023632067.1 | XP_023487835.1 | 827 | 85 | |||
| XM_023632068.1 | XP_023487836.1 | 821 | 85 | |||
| XM_023632069.1 | XP_023487837.1 | 763 | 85 | |||
| XM_023632070.1 | XP_023487838.1 | 826 | 85 | |||
| XM_023632071.1 | XP_023487839.1 | 756 | 85 | |||
| XM_023632072.1 | XP_023487840.1 | 691 | 85 | |||
| XM_023632073.1 | XP_023487841.1 | 684 | 85 | |||
| XM_023632074.1 | XP_023487842.1 | 954 | 85 | |||
| XM_023632075.1 | XP_023487843.1 | 1626 | 85 | |||
| XM_023632076.1 | XP_023487844.1 | 721 | 85 | |||
| XM_023632077.1 | XP_023487845.1 | 602 | 85 | |||
| XM_023632078.1 | XP_023487846.1 | 1747 | 85 | |||
| XM_023632079.1 | XP_023487847.1 | 1499 | 85 | |||
Set Membership: |
|