Search
Search
| Gene Symbol: NMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632251.1 | XP_023488019.1 | 1427 | 69 | ||
| XM_023632252.1 | XP_023488020.1 | 1265 | 69 | |||
| XM_023632253.1 | XP_023488021.1 | 1259 | 69 | |||
| XM_023632254.1 | XP_023488022.1 | 1377 | 69 | |||
Set Membership: |
|