Search
Search
| Gene Symbol: GDI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632340.1 | XP_023488108.1 | 1053 | 118 | ||
| XM_023632341.1 | XP_023488109.1 | 1110 | 118 | |||
| GenBank mRNA | HQ890256.1 | - | 3 - 4 | 345 | 118 | |
Set Membership: |
|