Search
Search

C3H16orf70| Gene Symbol: C3H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496846.6 | XP_001496896.2 | 959 | 87 | ||
| XM_005608323.3 | XP_005608380.1 | 967 | 87 | |||
| XM_005608325.3 | XP_005608382.1 | 963 | 87 | |||
| XM_014738377.2 | XP_014593863.1 | 889 | 87 | |||
| XM_014738378.2 | XP_014593864.1 | 963 | 87 | |||
| XM_014738379.2 | XP_014593865.1 | 969 | 87 | |||