Search
Search

NFATC3| Gene Symbol: NFATC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637170.1 | XP_023492938.1 | 1544 | 62 | ||
| XM_023637171.1 | XP_023492939.1 | 1544 | 62 | |||
| GenBank mRNA | JL629018.1 | - | 1 - 2 | 125 | 62 | |
Set Membership: |
|