Search
Search
| Gene Symbol: BDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499229.4 | XP_001499279.1 | 545 | 105 | ||
| XM_005608584.3 | XP_005608641.1 | 321 | 105 | |||
| XM_023637606.1 | XP_023493374.1 | 285 | 105 | |||
| GenBank mRNA | HQ890078.1 | - | 1 - 2 | 96 | 105 | |
Set Membership: |
|