Search
Search
| Gene Symbol: BDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499229.4 | XP_001499279.1 | 920 | 94 | ||
| XM_005608584.3 | XP_005608641.1 | 696 | 94 | |||
| XM_023637606.1 | XP_023493374.1 | 660 | 94 | |||
| GenBank mRNA | HQ890078.1 | - | 5 - 6 | 471 | 94 | |
Set Membership: |
|