Search
Search

SPARCL1| Gene Symbol: SPARCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496019.4 | XP_001496069.1 | 2400 | 123 | ||
| XM_014738731.2 | XP_014594217.2 | 2009 | 123 | |||
| XM_014738732.2 | XP_014594218.1 | 2305 | 123 | |||
Set Membership: |
|