Search
Search

CENPC| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497321.6 | XP_001497371.3 | 869 | 138 | ||
| XM_023638031.1 | XP_023493799.1 | 745 | 138 | |||
| XM_023638032.1 | XP_023493800.1 | 819 | 138 | |||
Set Membership: |
|