Search
Search

ADGRL3| Gene Symbol: ADGRL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638684.1 | XP_023494452.1 | 3284 | 69 | ||
| XM_023638685.1 | XP_023494453.1 | 3479 | 69 | |||
| XM_023638686.1 | XP_023494454.1 | 3353 | 69 | |||
| XM_023638687.1 | XP_023494455.1 | 3571 | 69 | |||
| XM_023638688.1 | XP_023494456.1 | 3410 | 69 | |||
| XM_023638689.1 | XP_023494457.1 | 3269 | 69 | |||
| XM_023638690.1 | XP_023494458.1 | 3269 | 69 | |||
| XM_023638691.1 | XP_023494459.1 | 3284 | 69 | |||
| XM_023638692.1 | XP_023494460.1 | 3284 | 69 | |||
| XM_023638693.1 | XP_023494461.1 | 3284 | 69 | |||
| XM_023638694.1 | XP_023494462.1 | 3203 | 69 | |||
| XM_023638695.1 | XP_023494463.1 | 3284 | 69 | |||
| XM_023638696.1 | XP_023494464.1 | 3284 | 69 | |||
| XM_023638697.1 | XP_023494465.1 | 3284 | 69 | |||
| XM_023638698.1 | XP_023494466.1 | 3080 | 69 | |||
| XM_023638699.1 | XP_023494467.1 | 3284 | 69 | |||
| XM_023638700.1 | XP_023494468.1 | 3065 | 69 | |||
| XM_023638701.1 | XP_023494469.1 | 3284 | 69 | |||
| XM_023638702.1 | XP_023494470.1 | 3284 | 69 | |||
| XM_023638703.1 | XP_023494471.1 | 3284 | 69 | |||
| XM_023638704.1 | XP_023494472.1 | 3284 | 69 | |||
Set Membership: |
|