Search
Search
| Gene Symbol: ARL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738870.2 | XP_014594356.1 | 670 | 63 | ||
| XM_014738871.2 | XP_014594357.1 | 720 | 63 | |||
| XM_023638054.1 | XP_023493822.1 | 749 | 63 | |||
Set Membership: |
|