Search
Search

DCUN1D4| Gene Symbol: DCUN1D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608780.3 | XP_005608837.2 | 419 | 85 | ||
| XM_005608781.3 | XP_005608838.1 | 540 | 85 | |||
| XM_023638104.1 | XP_023493872.1 | 509 | 85 | |||
| XM_023638105.1 | XP_023493873.1 | 415 | 85 | |||
| XM_023638106.1 | XP_023493874.1 | 546 | 85 | |||
| XM_023638107.1 | XP_023493875.1 | 509 | 85 | |||
| XM_023638108.1 | XP_023493876.1 | 509 | 85 | |||
| XM_023638109.1 | XP_023493877.1 | 325 | 85 | |||