Search
Search

BOD1L1| Gene Symbol: BOD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638344.1 | XP_023494112.1 | 687 | 89 | ||
| XM_023638345.1 | XP_023494113.1 | 687 | 89 | |||
| GenBank mRNA | JL617656.1 | - | 1 - 2 | 170 | 89 | |
Set Membership: |
|