Search
Search

SMYD3| Gene Symbol: SMYD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632601.1 | XP_023488369.1 | 1205 | 104 | ||
| XM_023632602.1 | XP_023488370.1 | 1154 | 104 | |||
| XM_023632603.1 | XP_023488371.1 | 1034 | 104 | |||
| XM_023632604.1 | XP_023488372.1 | 983 | 104 | |||
| GenBank mRNA | JL625420.1 | - | 7 - 8 | 688 | 104 | |
Set Membership: |
|