Search
Search

PPP1R12B| Gene Symbol: PPP1R12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632639.1 | XP_023488407.1 | 2216 | 64 | ||
| XM_023632640.1 | XP_023488408.1 | 2210 | 64 | |||
| XM_023632641.1 | XP_023488409.1 | 2219 | 64 | |||
| XM_023632642.1 | XP_023488410.1 | 2211 | 64 | |||
| XM_023632643.1 | XP_023488411.1 | 2215 | 64 | |||
| XM_023632644.1 | XP_023488412.1 | 2215 | 64 | |||
| XM_023632645.1 | XP_023488413.1 | 2028 | 64 | |||
| XM_023632646.1 | XP_023488414.1 | 2517 | 64 | |||
| XM_023632647.1 | XP_023488415.1 | 2217 | 64 | |||
| XM_023632648.1 | XP_023488416.1 | 2217 | 64 | |||
| XM_023632652.1 | XP_023488420.1 | 490 | 64 | |||
Set Membership: |
|