Search
Search

SEMA3C| Gene Symbol: SEMA3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638816.1 | XP_023494584.1 | 1496 | 102 | ||
| XM_023638817.1 | XP_023494585.1 | 1807 | 102 | |||
| XM_023638818.1 | XP_023494586.1 | 1611 | 102 | |||
Set Membership: |
|