Search
Search

KMT2E| Gene Symbol: KMT2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640149.1 | XP_023495917.1 | 804 | 65 | ||
| XM_023640150.1 | XP_023495918.1 | 804 | 65 | |||
| XM_023640151.1 | XP_023495919.1 | 658 | 65 | |||
| XM_023640153.1 | XP_023495921.1 | 364 | 65 | |||
Set Membership: |
|