Search
Search

KDM4B| Gene Symbol: KDM4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023644479.1 | XP_023500247.1 | 568 | 99 | ||
| XM_023644480.1 | XP_023500248.1 | 568 | 99 | |||
| XM_023644481.1 | XP_023500249.1 | 568 | 99 | |||
| XM_023644482.1 | XP_023500250.1 | 568 | 99 | |||
| XM_023644483.1 | XP_023500251.1 | 568 | 99 | |||
| XM_023644484.1 | XP_023500252.1 | 568 | 99 | |||
| XM_023644485.1 | XP_023500253.1 | 568 | 99 | |||
| XM_023644486.1 | XP_023500254.1 | 568 | 99 | |||
Set Membership: |
|