Search
Search

FOXRED1| Gene Symbol: FOXRED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502385.5 | XP_001502435.2 | 821 | 83 | ||
| XM_005611694.3 | XP_005611751.1 | 821 | 83 | |||
| XM_014741313.2 | XP_014596799.1 | 710 | 83 | |||
Set Membership: |
|