Search
Search

C7H11orf54| Gene Symbol: C7H11orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488169.6 | XP_001488219.1 | 400 | 71 | ||
| XM_014741730.2 | XP_014597216.1 | 402 | 71 | |||
| XM_014741731.2 | XP_014597217.1 | 366 | 71 | |||
| XM_023645563.1 | XP_023501331.1 | 326 | 71 | |||
Set Membership: |
|