Search
Search

ELAPOR2| Gene Symbol: ELAPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639101.1 | XP_023494869.1 | 1625 | 87 | ||
| XM_023639102.1 | XP_023494870.1 | 1626 | 87 | |||
| XM_023639103.1 | XP_023494871.1 | 1278 | 87 | |||
| XM_023639104.1 | XP_023494872.1 | 2488 | 87 | |||