Search
Search

GTPBP10| Gene Symbol: GTPBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490226.4 | XP_001490276.1 | 543 | 115 | ||
| XM_005609204.3 | XP_005609261.1 | 665 | 115 | |||
| XM_023639156.1 | XP_023494924.1 | 683 | 115 | |||
Set Membership: |
|