Search
Search

VPS50| Gene Symbol: VPS50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492642.5 | XP_001492692.1 | 1064 | 75 | ||
| XM_023639201.1 | XP_023494969.1 | 1083 | 75 | |||
| XM_023639202.1 | XP_023494970.1 | 1105 | 75 | |||
| XM_023639203.1 | XP_023494971.1 | 1419 | 75 | |||
| XM_023639204.1 | XP_023494972.1 | 921 | 75 | |||