Search
Search
| Gene Symbol: ASB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493628.5 | XP_001493678.1 | 2252 | 103 | ||
| XM_023639234.1 | XP_023495002.1 | 1226 | 103 | |||
| XM_023639236.1 | XP_023495004.1 | 1089 | 103 | |||
| XM_023639237.1 | XP_023495005.1 | 1140 | 103 | |||
Set Membership: |
|