Search
Search
| Gene Symbol: TAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609230.3 | XP_005609287.1 | 591 | 70 | ||
| XM_023639248.1 | XP_023495016.1 | 626 | 70 | |||
| XM_023639249.1 | XP_023495017.1 | 572 | 70 | |||
Set Membership: |
|