Search
Search

CALD1| Gene Symbol: CALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609422.3 | XP_005609479.1 | 1949 | 64 | ||
| XM_014739289.2 | XP_014594775.1 | 2793 | 64 | |||
| XM_014739291.2 | XP_014594777.1 | 2692 | 64 | |||
| XM_014739292.2 | XP_014594778.1 | 2414 | 64 | |||
| XM_014739293.2 | XP_014594779.1 | 2792 | 64 | |||
| XM_014739294.2 | XP_014594780.1 | 2790 | 64 | |||
| XM_014739295.2 | XP_014594781.1 | 2789 | 64 | |||
| XM_014739296.2 | XP_014594782.1 | 2510 | 64 | |||
Set Membership: |
|