Search
Search
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494787.4 | XP_001494837.1 | 649 | 85 | ||
| XM_014742048.2 | XP_014597534.1 | 650 | 85 | |||
| XM_023646845.1 | XP_023502613.1 | 675 | 85 | |||