Search
Search

ACACB| Gene Symbol: ACACB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496930.4 | XP_001496980.2 | 3762 | 61 | ||
| XM_005612493.3 | XP_005612550.2 | 3289 | 61 | |||
| XM_023646975.1 | XP_023502743.1 | 4086 | 61 | |||
| XM_023646976.1 | XP_023502744.1 | 3768 | 61 | |||
| XM_023646977.1 | XP_023502745.1 | 3737 | 61 | |||
Set Membership: |
|